Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DISP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AW0
10801-142EA 518 USD InStock
10801-142
Anti-DISP1 Rabbit Polyclonal Antibody
Antibodies
DISP1 is the mammalian homolog of the Drosophila Dispatched segment-polarity gene and a key regulator of the Hedgehog (Hh) signaling pathway during embryonic development. DISP1 is required to move Shh from site of synthesis during embryogenesis; mutations in the gene result in lethality at mid-gestation and prevents specification of ventral cell types in the neural tube. Recent results have shown that DISP1 mediates the basolateral secretion of Shh and regulates growth of mammalian long bones through the control of Ihh.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 2.5 ?g/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: DISP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty