Would you like to log in again?
DNA Ligase T4
Supplier: Rockland Immunochemicals
MB-106-0500 MB-106-0100
10801-156EA 284.89 USD InStock
10801-156 10801-154
DNA Ligase T4
Nucleic Acid Reagents Cloning Kits and Ligation
DNA Ligase T4 Molecular Biology Grade
  • 50 mM Tris-HCl, 10 mM MgCl2, 10 mM DTT, 1 mM ATP, pH 7.5 @ 25°C.
  • Ligation of nicks in dsDNA and joining dsDNA fragments with complementary overhangs > 2 bases in length.
Catalyzes the formation of a phosphodiester bond between juxtaposed 5' phosphate and 3' hydroxyl termini in duplex DNA or RNA. This enzyme will join blunt end and cohesive end termini as well as repair single stranded nicks in duplex DNA and some DNA/RNA hybrids.
One unit will catalyze the exchange of one nanomole 32-P from pyrophosphate into a Norit-absorbable compound in 20 minutes at 37° C (Weiss, B., et al., JBC 243, 4543 (1968).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty