Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DTX4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AY1
10801-198EA 518 USD InStock
10801-198
Anti-DTX4 Rabbit Polyclonal Antibody
Antibodies
The Deltex 4 E3 ubiquitin ligase (DTX4) is a member of a protein family that is involved in Notch signaling and neurogenesis (1). Recent studies have shown that DTX4 is also associated with the innate immune response (2). Following the activation of type I interferon signaling, the Nod-like receptor protein 4 (NLRP4) recruits DTX4 for Lys48-linked polyubiquitination and degradation of the kinase TBK1 (NAK), thereby maintaining immune homeostasis during antiviral innate immunity (2).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: DTX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty