Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FAIM Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BD0
10801-330EA 518 USD InStock
10801-330
Anti-FAIM Rabbit Polyclonal Antibody
Antibodies
The susceptibility of primary splenic B cells to Fas-mediated apoptosis is regulated in a receptor-specific fashion. Terminal effectors of B cell Fas-resistance include the known anti-apoptotic proteins Bcl-xL, FLIP, and a recently identified protein termed FAIM. This molecule is broadly expressed in various tissues and exists in at least three isoforms. It is thought that resistance to Fas killing via increased expression of FAIM protects foreign antigen-specific B cells during interactions with FasL-bearing T cells whereas autoreactive B cells are deleted via Fas-dependent cytotoxicity. More recent results have indicated that FAIM interacts with both Trk and p75 neurotrophin receptor and may play a role in promoting neurite outgrowth in different neuronal systems by a mechanism involving the activation of NF-kB and the Ras-ERK pathway.

Recommended Dilutions: ELISA: 1:50,000-1:100,000; Western Blot: 5-10 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: FAIM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty