Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FAIM2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BD1
10801-332EA 518 USD InStock
10801-332
Anti-FAIM2 Rabbit Polyclonal Antibody
Antibodies
Programmed cell death regulates a number of biological processes such as normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer. FAIM2 is a recently identified protein that can inhibit the apoptotic signal transduced by the Fas receptor but not from the related tumor necrosis factor-alpha death signal. In this respect, FAIM2 is functionally similar to the anti-apoptotic proteins FAIM, FLIP and Bcl-xL. FAIM2, a seven membrane spanning protein, can bind the Fas receptor but does not regulate Fas expression or inhibit binding of FADD to Fas. FAIM2 is widely distributed, but highly expressed in the hippocampus and other neural tissues. FAIM2 was also identified as the neural membrane protein 35 (NMP35) and its expression is known to be regulated by the Phosphatidylinositol 3-kinase-Akt/PKB pathway.

Recommended Dilutions: ELISA: 1:50,000; Immunohistochemsitry: 5 ?g/mL; Western Blot: 5 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: FAIM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty