Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FKBP1A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BE8
10801-420EA 518 USD InStock
10801-420
Anti-FKBP1A Rabbit Polyclonal Antibody
Antibodies
FKBP, or FK506 binding protein, is a family of proteins that have prolyl isomerase activity and are related to the cyclophilins in function (1). FKBP12, also called FKBP1, was originally identified as a target of FK506 and rapamycin activity and catalyzes the transition between cis and transproline residues critical for proper folding of proteins (2). FKBP12 is an abundant, evolutionarily conserved cytoplasmic protein that is an important regulator of diverse array of cellular processes including T cell activation, entry into the cell cycle and intracellular calcium release (3-5).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: FKBP1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty