Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FKBP15 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BE9
10801-422EA 518 USD InStock
10801-422
Anti-FKBP15 Rabbit Polyclonal Antibody
Antibodies
FKBP15, also known as FKBP133, is a member of the FK506-binding protein family, a group of proteins initially identified as immunophilins, targets for the immunosupressant drugs FK506 and Rapamycin. FKBP15 is expressed in the developing nervous system and contains a domain similar to Wiskott-Aldrich syndrome protein homology region 1 (WH1) in addition to the FK506-binding protein motif. FKBP15 is distributed along the axonal shafts and partially co-localizes with F-actin in the growth cones of dorsal root ganglion neurons; overexpression of FKBP15 resulted in the number of filopodia in transfected neurons, suggesting that FKBP15 modulates growth cone behavior. FKBP15 has also been shown to associate with both microtubules and the actin filament systems and disruption of its expression by RNAi resulted in delayed transport of early endosomes in HeLa cells indicating that FKBP15 is also involved in the transport of early endosomes. At least three isoforms of FKBP15 are known to exist.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 2.5 ?g/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: FKBP15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty