Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FOXA1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BG0
10801-456EA 518 USD InStock
10801-456
Anti-FOXA1 Rabbit Polyclonal Antibody
Antibodies
FOXA1 is one of three members of the FOXA family, a subset of the forkhead family of transcription factors which play vital roles in development. It has also been implicated in the development of a number of other organs including the androgen and estrogen regulated tissues of the breast and prostate. FOXA1 modulates the transcriptional activity of nuclear hormone receptors and regulates apoptosis by inhibiting the expression of BCL-2. It is an essential protein for the transcriptional activity of both androgen receptor (AR) and estrogen receptor-a (ER). FOXA1 plays a pivotal role from early stage cancer through to drug resistant and metastatic disease. FOXA1 is not only an attractive therapeutic target but could potentially function as a novel biomarker.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: FOXA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty