Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FOXA2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BG1
10801-458EA 518 USD InStock
10801-458
Anti-FOXA2 Rabbit Polyclonal Antibody
Antibodies
FOXA2 is one of three members of the FOXA family, a subset of the forkhead family of transcription factors which play vital roles in development. FOXA2 was initially identified through library screening as a closely related homolog of FOXA1. Both FOXA2 and FOXA1 act as transcriptional activators in adult liver and also play a role in body axis formation, neural tube patterning and definitive endoderm formation during gastrulation.

Recommended Dilutions: ELISA: 1:10,000; Immunofluorescence Microscopy: User Optimized; Western Blot: 1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: FOXA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty