Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FOXG1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BG3
10801-462EA 518 USD InStock
10801-462
Anti-FOXG1 Rabbit Polyclonal Antibody
Antibodies
FOXG1 belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain and plays an important role in the development of the brain and telencephalon (1). It is essential for the proliferation of progenitor cells in the cerebral cortex and influences regional patterning in the mammalian telencephalon (2,3). Expression is restricted to the neurons of the developing telencephalon (3). The loss of FOXG1 results in an accelerated rate of neuronal differentiation and the shortening of the neurogenetic period in the embryonic cerebral cortex (2,4).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: FOXG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty