Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FOXO1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BG5
10801-466EA 518 USD InStock
10801-466
Anti-FOXO1 Rabbit Polyclonal Antibody
Antibodies
FOXO1, also known as FKHR or ForkHead in Rhabdomyosarcoma, is a 70 kDa protein which is a ubiquitously expressed member of a subfamily of the forkhead homeotic gene family of transcription factors and shuttles between the cytoplasm and nucleus. FOXO transcription factors are key players of cell fate decisions, metabolism, stress resistance, tumor suppression and are regulated by growth factors, oxidative stress or nutrient deprivation. In insulin-responsive tissues, stress or nutrient abundance triggers phosphorylation by PKB/AKT, blocking nuclear translocation and activity. Genetic mutations involving FOXO1A are a cause of alveolar rhabdomyosarcoma (RMS2). Recent studies link the anti-tumor activity of FOXO1 and the process of autophagy.

Recommended Dilutions: ELISA: 1:10,000; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: FOXO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty