Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FREM2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BH2
10801-488EA 518 USD InStock
10801-488
Anti-FREM2 Rabbit Polyclonal Antibody
Antibodies
FREM2 is a member of the FRAS1-related extracellular matrix protein family and like FREM1, mutations in the FREM2 gene are associated with Fraser syndrome. FREM2 localizes to the basement membrane, forming a ternary complex that plays a role in epidermal-dermal interactions during morphogenetic processes and is thought to be required for maintaining the integrity of the skin epithelium and the differentiated state of renal epithelia. The FREM2 gene is one of several genes whose transcription is affecteded by TFAP2C, a transcription factor involved in mammary development, differentiation, and oncogenesis.

Recommended Dilutions: ELISA: User Optimized; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: User Optimized; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: FREM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty