Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FRMPD1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BH4
10801-496EA 518 USD InStock
10801-496
Anti-FRMPD1 Rabbit Polyclonal Antibody
Antibodies
The FERM and PDZ domain containing (FRMPD) protein family consists of four proteins that contain a FERM (Four-point-one, erzin, radixin, moesin) domain and at least one PDZ (PSD-95/Discs large/Zonula-occuldens-1) domain. FRMPD1 was initially identified through a yeast two-hybrid screening with Activator of G-protein signaling 3 (AGS3), a protein that acts to stabilize the GDP-bound conformation of Gai, as the bait. FRMPD1 binds to the tetratricopeptide repeat of AGS3 and stabilizes AGS3 in a membrane environment. Suppression of FRMPD1 in Cath.a-differentiated neuronal cells decreased the level of endogenous AGS3 in membrane fractions by ~50% and enhanced the inhibition of forskolin-induced increases in cAMP. Co-immunoprecipitation studies indicate that the interaction of AGS3 with FRMPD1 is mutually exclusive with the interaction of AGS3 with Gai3, suggesting that FRMPD1 may position AGS3 in the membrane where it can then interact with Galphai.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: FRMPD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty