Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FXYD7 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BH9
10801-510EA 518 USD InStock
10801-510
Anti-FXYD7 Rabbit Polyclonal Antibody
Antibodies
FXYD7 is a member of a family of small membrane proteins that share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD and containing seven invariant and six highly conserved amino acids. The FXYD proteins are tissue-specific regulators of Na, K-ATPase, with FXYD7 initially identified as a brain-specific member. FXYD7 interacts with Na, K-ATPase through its transmembrane domain and is thought to influence the affinity of Na, K-ATPase for external K+ and Na+ ions. Other members of the FXDY family have similar functions: FXYD2 regulates the properties of Na, K-ATPase, while FXYD1 (phospholemman), FXYD3 (MAT-8), FXYD4 (CHIF), and FXYD5 (RIC) have been shown to induce channel activity in experimental expression systems.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 2.5 ?g/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: FXYD7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty