Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GABARAPL2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BJ1
10801-522EA 518 USD InStock
10801-522
Anti-GABARAPL2 Rabbit Polyclonal Antibody
Antibodies
Gamma-aminobutyric acid (GABA) is the main inhibitory transmitter by increasing a Cl-conductance that inhibits neuronal firing in the central nervous system (1). It has been shown to activate both ionotropic (GABAA) and metabotropic (GABAB) receptors as well as a third class of receptors called GABAC (2). GABARAPL2 (GABAA receptor-associated protein-like 2), also known as GATE16, was initially identified as a membrane transport modulator and is a mammalian ortholog to the autophagy protein ATG8 (3,4). It is thought that GABARAPL2 and other members of the ATG8 family act as scaffolds for assembly of the Unc-51 like kinase (ULK) complex in the formation of autophagosomes (5).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 ?g/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: GABARAPL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty