Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GDNF Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-R08
10801-620EA 487 USD InStock
10801-620
Anti-GDNF Rabbit Polyclonal Antibody
Antibodies
GDNF, Glial cell-derived neurotrophic factor, is a protein that, in humans, is encoded by the GDNF gene. GDNF is a small protein that potently promotes the survival of many types of neurons. GDNF is a founding member of the GDNF family of ligands (GFL).The GDNF gene is mapped to human chromosome 5p13.1-p12 by fluorescence in situ hybridization (FISH). This gene encodes a highly conserved neurotrophic factor. The recombinant form of this protein was shown to promote the survival and differentiation of dopaminergic neurons in culture, and was able to prevent apoptosisof motor neurons induced by axotomy. The encoded protein is processed to a mature secreted form that exists as a homodimer. The mature form of the protein is a ligand for the product of the RET (rearranged during transfection) protooncogene. This antibody is suitable for researchers interested in neuroscience research.

Recommended Dilutions: Immunohistochemsitry: 1:100-1:500; Western Blot: 0.5µg/mL; 0.05mg Thimerosal

Type: Primary
Antigen: GDNF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty