Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GSN Mouse Monoclonal Antibody [clone: GEL-42]

Supplier: Rockland Immunochemicals
200-301-P37
10801-624EA 528 USD InStock
10801-624
Anti-GSN Mouse Monoclonal Antibody [clone: GEL-42]
Antibodies
Gelsolin, a protein of leukocytes, platelets, and other cells, severs actin filaments in the presence of submicromolar calcium, thereby solating cytoplasmic actin gels. A gelsolin variant with 23 more N-terminal amino acids is a plasma component probably involved in the clearance of actin, the most abundant human protein, from the circulation. Gelsolin is located in 9q34. Plasma and cytoplasmic gelsolins are encoded by a single gene and contain a duplicated actin-binding domain. This antibody is suitable for researchers interested in cell research and cytoskeletal signaling research.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 1:100-1:500; Western Blot: 0.5-1µg/ml; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: GSN
Clonality: Monoclonal
Clone: GEL-42
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rabbit Proteins.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty