Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GFAP Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S206B-9]

Supplier: Rockland Immunochemicals
200-308-W56
10801-638EA 539 USD InStock
10801-638
Anti-GFAP Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S206B-9]
Antibodies
The 50 kDa type III intermediate filament protein glial fibrillary acidic protein (GFAP) is a major structural component of astrocytes. GFAP associates with the calcium binding protein annexin II-p2 and S-100. Association with these proteins together with phosphorylation regulates GFAP polymerization. Astroycytes respond to brain injury by proliferatin (astrogliosis), and one of the first events to occur during astrocyte proiliferation is increased GFAP expression. Interestingly, antibodies to GFAP have been detected in individuals with dementia. Anti-GFAP is ideal for investigators involved in Neuroscience Research, including Alexander Disease, Oligodendroglioma, Cytoskleton Remolding Neurofilaments and PIP3/AKT Signaling.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: User Optimized; Western Blot: 1:1000; 0.1% (w/v) Sodium Azide

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: S206B-9
Conjugation: RPE (R-Phycoerythrin)
Epitope: C-terminal
Host: Mouse
Isotype:
Reactivity: Human, Rat, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty