Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACVR1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X66 600-401-X66
10801-658EA 518 USD InStock
CA10801-658 10801-658
Anti-ACVR1 Rabbit Polyclonal Antibody
Antibodies
Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I and two type II receptors. Unlike ACVR1B and ACVR1C, ACVR1, also known as activin receptor-like kinase 2 (ALK2), can not transduce activin-mediated signaling, but will transduce BMP and Mullerian inhibiting substance (MIS) group signaling. It is thought that ACVR1 also inhibits activin signaling by blocking the binding of activin to its type II receptor. Recent studies indicate that genetic variation in ACVR1 is associated with polycystic ovary syndrome, suggesting that ACVR1 signaling contributes to disturbed folliculogenesis in these patients.

Recommended Dilutions: ELISA: 1:10,000; Western Blot: 1 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: ACVR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty