Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GFP Mouse Monoclonal Antibody [clone: 9F9.F9]

Supplier: Rockland Immunochemicals
600-301-215S
10801-682EA 145 USD InStock
10801-682
Anti-GFP Mouse Monoclonal Antibody [clone: 9F9.F9]
Antibodies
Mouse anti-GFP antibody is functional by western blot, ELISA, Immunofluorescene Micorscopy and Immunohistochemistry. Green fluorescent protein is a 27 kDa protein produced from the jellyfish Aequorea victoria, which emits green light (emission peak at a wavelength of 509nm) when excited by blue light. GFP is an important tool in cell biology research. GFP is widely used enabling researchers to visualize and localize GFP-tagged proteins within living cells without the need for chemical staining.

Recommended Dilutions: ELISA: 1:80,000 - 1:500,000; Flow Cytometry: User Optimized; Immunohistochemsitry: 1:1,000 - 1:5,000; Immunofluorescence Microscopy: User Optimized; Western Blot: 1:300 - 1:30,000; contains contains 0.01% (w/v) Sodium Azide

Type: Tag
Antigen: Green Fluorescent Protein (GFP) fusion protein
Clonality: Monoclonal
Clone: 9F9.F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty