Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GJB2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BL1
10801-696EA 518 USD InStock
10801-696
Anti-GJB2 Rabbit Polyclonal Antibody
Antibodies
The Gap junction beta-2 protein (GJB2), also known as Connexin 26, is member of the gap junction protein family which form structures that were shown to consist of cell-to-cell channels that facilitate the transfer of ions and small molecules between cells (1). Mutations in the GJB2 gene are thought to be responsible for as much as 35-45% of congenital sensorineural hearing loss in some populations (2). Other mutations in this gene have also been linked to a wide array of skin diseases (3).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: GJB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty