Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GLIPR1L1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BL5
10801-728EA 518 USD InStock
10801-728
Anti-GLIPR1L1 Rabbit Polyclonal Antibody
Antibodies
The GLIPR1-like 1 protein (GLIPR1L1) gene is part of a p53 target gene cluster that includes the related proteins GLIPR1 and GLIPR1L2 (1). GLIPR1L1 is similar to both the pathogenesis-related protein (PR) superfamily and the cysteine-rich secretory protein (CRISP) family (2). GLIPR1 is a tumor suppressor whose expression is regulated by p53 (3). Unlike GLIPR1, GLIPR1L1 is expressed primarily in the testis and is thought to be involved in the binding of sperm to the oocyte complex (4).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: 5 ?g/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: GLIPR1L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty