Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GLIS2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BL7
10801-732EA 518 USD InStock
10801-732
Anti-GLIS2 Rabbit Polyclonal Antibody
Antibodies
GLIS2, also known as neuronal Krueppel-like, is a 524 amino acid protein that belongs to the GLI C2H2-type zinc-finger protein family (1). GLIS2 can act either as a transcription repressor or as a transcription activator and may be involved in neuron differentiation (1,2). GLIS2 is expressed at high levels in kidney and at low levels in heart, lung and placenta. Mutations of GLIS2 have been suggested to be associated with development of progressive chronic kidney disease with characteristics resembling nephronophthisis (3,4).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: GLIS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty