Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ARHGAP39 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-Y59 600-401-Y59
10801-906EA 518 USD InStock
CA10801-906 10801-906
Anti-ARHGAP39 Rabbit Polyclonal Antibody
Antibodies
Rho GTPases are important regulators of the actin cytoskeleton and consequently influence the shape and migration of cells. GTPase-activating proteins (GAPs) accelerate the intrinsic rate of GTP hydrolysis of Ras-related proteins. ARHGAP39 (Rho GTPase activating protein 39) is a 1, 083 amino acid nuclear protein that contains one MyTH4 domain, one Rho GAP domain and two WW domains. It is encoded by a gene located on human chromosome 8, which is associated with a variety of diseases and malignancies. ARHGAP39 regulates robo signaling and plays important roles in axon guidance.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1 - 2 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: ARHGAP39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty