Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ARR3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-Y62 600-401-Y62
10801-914EA 518 USD InStock
CA10801-914 10801-914
Anti-ARR3 Rabbit Polyclonal Antibody
Antibodies
Arrestin 3 (ARR3) belongs to the Arrestin family of proteins that function as negative regulators of G protein-coupled receptor (GPCR) signaling (1,2). ARR3 binds the c-Jun N-terminal kinase 3 (JNK3) and scaffolds the apoptosis signal-regulating kinase 1 (ASK1)-MAPK kinase4-JNK3 cascade in a receptor-independent fashion, promoting JNK3 phosphorylation (3,4). ARR3 also mediates the internalization of the C-C chemokine receptor CCR7 upon the binding of its ligand CCL19, but not CCL21, suggesting that it helps mediate CCR7 receptor desensitization (5).

Recommended Dilutions: ELISA: 1:5000-1:10000; Western Blot: 0.5 - 1 µg/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: ARR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty