Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BCAS3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-Z40
10802-032EA 518 USD InStock
10802-032
Anti-BCAS3 Rabbit Polyclonal Antibody
Antibodies
The Breast carcinoma amplified sequence (BCAS3) gene is localized to 17q23, a region that is often amplified in breast cancers. Copy number gains at this region have also been reported in other tumors such as brain, lung, liver, testis and bladder. BCAS3 is thought to be an estrogen receptor (ER)-alpha co-activator that acts through PELP1, another ER-alpha that in turn activates BCAS3 expression. This may set up a positive feedback loop leading to ER-a signal amplification in the cell which may play a significant role in breast cancer. BCAS3 expression has also been observed in embryonic stem cells and vascular precursors, suggesting that it may also play a role in embryogenesis and tumor angiogenesis.

Recommended Dilutions: ELISA: 1:5000-1:10000; Immunohistochemsitry: 5 µg/mL; Immunofluorescence Microscopy: 20 µg/mL; Western Blot: 0.5 - 1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: BCAS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty