Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BECN1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-W96
10802-050EA 518 USD InStock
10802-050
Anti-BECN1 Rabbit Polyclonal Antibody
Antibodies
Beclin-1 plays a key role in tumor suppression and autophagy. It has been suggested that autophagy proteins such as beclin-1 can inhibit tumorigenesis and that decreased expression of these proteins may contribute to the development of human malignancies. Beclin-1 has been shown to interact with 14-3-3 proteins through Akt-mediated phosphorylation at Ser234. Beclin 1 pS234 Antibody is ideal for researchers interested in cancer research.

Recommended Dilutions: ELISA: 1:2,500; Western Blot: 1:250;

Type: Primary
Antigen: BECN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Bovine, Canine, Mouse, Non-Human Primates, Rat, Sheep

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty