Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EMX1 Chicken Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-901-BA0
10802-090EA 518 USD InStock
10802-090
Anti-EMX1 Chicken Polyclonal Antibody
Antibodies
EMX1 is a homeobox transcription factor involved in specifying cell fates in the developing central nervous system and it participates in the development of olfactory neurons. EMX1 is specifically expressed in the developing telencephalic cortex expression, where epxression is restricted to pyramidal neurons. EMX1 is a reliable marker of pyramidal neurons and pyramidal cell lineage. EMX1 has been shown to be one of the downstream target genes for Gli-Kruppel family member 3 (Gli3) transcription factor, which is a part of the Sonic hedgehog-Patched-Gli signaling pathway important in endocrine signaling.

Recommended Dilutions: ELISA: 1:10,000; Western Blot: 1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: EMX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty