Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RAPGEF3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BB1
10802-100EA 518 USD InStock
10802-100
Anti-RAPGEF3 Rabbit Polyclonal Antibody
Antibodies
EPAC1, also known as Rap guanine nuclear exchange factor 3 and cAMPGEF-I, is widely expressed but most prominently in brain, heart, kidney, pancreas, spleen, ovary, thyroid and spinal cord. EPAC1 is a cAMP-binding protein with intrinsic guanine nuclear exchange factor activity that couples cAMP production to the activation of Rap, a GTPase belonging to the Ras family. This activation of Rap influences numerous cellular processes such as integrin-mediated cell adhesion, vascular endothelial barrier formation, and cardiac myocyte gap junction formation. Recently, EPAC1 has been suggested to also be involved in the cAMP-dependent regulation of ion channel formation, intracellular Ca++ signalling, ion transporter activity, and exocytosis.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 2.5 ?g/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: RAPGEF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty