Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EPM2A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BB4
10802-110EA 518 USD InStock
10802-110
Anti-EPM2A Rabbit Polyclonal Antibody
Antibodies
The Epilepsy, progressive myoclonus type 2A protein (EPM2A) is a dual-specificity phosphatase that associates with polyribosomes. Mutations in this gene have been associated with myoclonic epilepsy of Lafora. EPM2A interacts with a number of proteins known to be involved in glycogen metabolism and has been shown to have robust phosphatase activity against a phosphorylated complex carbohydrate, suggesting that EPM2A may be involved in the regulation of glycogen metabolism.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunofluorescence Microscopy: User Optimized; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: EPM2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty