Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ERAP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BB5
10802-112EA 518 USD InStock
10802-112
Anti-ERAP1 Rabbit Polyclonal Antibody
Antibodies
The endoplasmic reticulum (ER) aminopeptidase 1 (ERAP1), a member of the peptidase M1 family, plays a central role in peptide trimming, a step required for the generation of most HLA class I-binding peptides (1,2). It is also designated as adipocyte-derived leucine aminopeptidase (A-LAP), puromycin-insensitive leucine-specific aminopeptidase (PILS-AP) and aminopeptidase regulator of TNFR1 shedding (ARTS-1) (3). ERAP1 is localized to the lumen of the ER and induced by interferon. It may be involved in the regulation of blood pressure through the inactivation of angiotensin II and/or the generation of bradykinin in the kidney (3,4).

Recommended Dilutions: ELISA: User Optimized; Immunohistochemsitry: User Optimized; Immunofluorescence Microscopy: User Optimized; Western Blot: User Optimized; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: ERAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty