Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ERAP2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BB6
10802-114EA 518 USD InStock
10802-114
Anti-ERAP2 Rabbit Polyclonal Antibody
Antibodies
The endoplasmic reticulum (ER) aminopeptidase 2 (ERAP2), a member of the peptidase M1 family, like the related protein ERAP1, plays a central role in peptide trimming, a step required for the generation of most HLA class I-binding peptides (1,2). Like ERAP1, ERAP2 is localized to the lumen of the ER and is thought to associate with ERAP1 as a heterodimer (1). Both ERAP1 and ERAP2 have been linked to several human diseases ranging from infections to autoimmunity and cancer, and may play a role in the innate immune response (reviewed in 3).

Recommended Dilutions: ELISA: User Optimized; Western Blot: User Optimized; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: ERAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty