Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BrdU Mouse Monoclonal Antibody [clone: 29G6.E8]

Supplier: Rockland Immunochemicals
200-301-H50
10802-262EA 540 USD InStock
10802-262
Anti-BrdU Mouse Monoclonal Antibody [clone: 29G6.E8]
Antibodies
Bromodeoxyuridine (5-bromo-2'-deoxyuridine, BrdU) is a synthetic thymidine nucleoside analog. BrdU is commonly used to allow the detection of growing or proliferating cells in living tissues. During the S-phase of cell division, DNA replication occurs, and BrdU can be incorporated into the newly synthesized DNA by substituting for naturally occurring thymidine. Antibodies specific for BrdU are subsequently used to detect the incorporated BrdU thymidine analog. This highlights cells that were actively replicating their DNA and is suggestive of actively growing cells. Antibody binding usually requires the DNA to be denatured, typically by exposing the cells to acid or heat.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Flow Cytometry: 1:50-1:100; Immunohistochemsitry: 1:100-1:500; Immunofluorescence Microscopy: 1:500-1:3000; Western Blot: 1:2000 - 1:5000; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: BrdU
Clonality: Monoclonal
Clone: 29G6.E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: 0

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty