Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CACNA1H Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AA3
10802-272EA 518 USD InStock
10802-272
Anti-CACNA1H Rabbit Polyclonal Antibody
Antibodies
The calcium channel protein CACNA1H is a T-type member of the alpha-1 subunit family that is part of the voltage-dependent calcium channel complex which mediates the influx of calcium ions into the cell upon membrane polarization (1,2). CACNA1H is a subunit of Cav3.2 T-type calcium channel that is involved in neurological disorders such as epilepsy and pain (2,3). CACNA1H associates with the caveolin protein Cav-3 and it is thought that Cav-3 regulates the Protein Kinase A (PKA) modulation of CACNA1H-containing Cav3.2 T-type calcium channels (4).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CACNA1H
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty