Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CHOX_ALCSP Goat Polyclonal Antibody (BAC)

Supplier: Rockland Immunochemicals
200-106-094-0100
10802-380EA 498 USD InStock
10802-380
Anti-CHOX_ALCSP Goat Polyclonal Antibody (BAC)
Antibodies
Choline Oxidase is a flavoprotein responsible for the catalyzing the conversion of choline to betaine aldehyde in animals and bacteria. Anti-Choline Oxidase antibody is ideal for researchers interested in Metabolism and Signal Transduction research.

Anti-Choline Oxidase has been assayed against 1.0 ug of Choline Oxidase in a standard capture ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature.

Type: Primary
Antigen: CHOX_ALCSP
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Goat
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty