Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TRAF3IP2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AK9
10802-392EA 518 USD InStock
10802-392
Anti-TRAF3IP2 Rabbit Polyclonal Antibody
Antibodies
Nuclear factor kappa B (NF-kB) is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-kB mediates the expression of a great variety of genes in response to extracellular stimuli. NF-kB associates with IkB proteins in the cell cytoplasm, which inhibit NF-kB activity. IkB is phosphorylated by IkB kinase (IKK) complex that contains IKKa, IKKb, and IKKgamma. A novel molecule that associates with and activates IKK was recently identified and designated CIKS (for connection to IKK and SAPK/JNK) and Act1 (for NF-kB activator 1). CIKS directly interacts with IKKgamma. CIKS/Act1 also activates activating transcription factor (ATF) and activator protein 1 (AP-1) through Jun kinase (JNK). These results indicate that CIKS/Act1 is involved in the inflammation and stress responses. CIKS/Act1 is ubiquitously expressed in human tissues.

Recommended Dilutions: ELISA: 1:10,000; Immunohistochemsitry: 5 µg/mL; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: TRAF3IP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty