Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CNOT4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-AM2
10802-414EA 518 USD InStock
10802-414
Anti-CNOT4 Rabbit Polyclonal Antibody
Antibodies
CNOT4 is a component of the CCR4-NOT transcription complex, a complex that is implicated in the repression of RNA polymerase II transcription. In the CCR4-NOT complex, CNOT4 acts as an E3 ubiquitin-protein ligase and interacts with a subset of E2 ubiquitin-conjugating enzymes through a unique C4C4 RING domain. This E3 ligase activity was shown to be dependent on the selective and specific interaction with the ubiquitin conjugating enzyme UbcH5B. In yeast, mutations in CNOT4 that prevented its interaction with the UbcH5B homolog UBC4 caused increased sensitivity to hydroxyurea, heat shock, and hygromycin B, suggesting that CNOT4 and UbcH5B are involved in stress response in vivo. Multiple isoforms of CNOT4 are known to exist.

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: CNOT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty