Would you like to log in again?
Donkey Plasma Sterile In Sodium Heparin, Rockland Immunochemicals
Supplier: Rockland Immunochemicals
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
D520-06-0050 D520-06-0100
10802-524EA 739 USD InStock
10802-524 10802-526
Donkey Plasma Sterile In Sodium Heparin, Rockland Immunochemicals
Blood
Sterile Plasma in Anticoagulant
  • 0.22 micron filter and found to be negative for bacterial growth after incubation on trypticase soy agar for 24 hours, 48 hours, and 72 hours
  • prepared by collection of whole blood into said anticoagulant
  • Red blood cells are removed through a process of centrifugation and filtration techniques - leaving non-sterile plasma behind
used as a component of bioassays, immunoassays or enzyme assays
used as a supplement to cell culture media
provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health
Be certain to maintain Good Cell Culture Practice, and maintain sterility of cultures that require media supplementation.
pH: normal
Immunoelectrophoresis: normal
Hemoglobin: normal
IgG Concentration: normal
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty