Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DCX Mouse Monoclonal Antibody [clone: 30]

Supplier: Rockland Immunochemicals
200-301-X04
10802-538EA 528 USD InStock
10802-538
Anti-DCX Mouse Monoclonal Antibody [clone: 30]
Antibodies
Doublecortin, or DCX, is a microtubule associated protein that is expressed almost exclusively in very early neuronal development, making it an excellent marker for developing neuronal cells. Defects in the DCX gene lead to X-linked lissencephaly which is characterized by a lack of normal folds on the surface of the brain resulting in a smooth cerebral cortex caused by abnormal migration of neurons during development. Doublecortin Antibody is ideal for researchers interested in neuroscience research.

Recommended Dilutions: ELISA: 1:50,000; Immunofluorescence Microscopy: 1:1000; Western Blot: 1:5000; 0.1% (w/v) Sodium Azide

Type: Primary
Antigen: DCX
Clonality: Monoclonal
Clone: 30
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Bovine, Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty