Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EEF1A2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X05
10802-574EA 518 USD InStock
10802-574
Anti-EEF1A2 Rabbit Polyclonal Antibody
Antibodies
Eukaryotic Elongation Factor eEF1A exists in two variant forms, eEF1A1 and eEF1A2. While eEF1A1 is almost ubiquitously expressed in humans, eEF1A2 is predominantly found in heart, brain, and skeletal muscle. Expression of eEF1A2 may have a role in ovarian cancer, as its expression is drastically increased in human ovarian tumors. Due to differences in structural models between the two isoforms, eEF1A1 and eEF1A2 likely have variant-specific phosphorylation sites. Ribosome-associated JNK phosphorylates Ser358 on eEF1A2 to promote degradation of newly synthesized polypeptides by the proteasome. eEF1A2 pS358 Antibody is ideal for researchers interested in cancer research.

Recommended Dilutions: ELISA: 1:10,000; Western Blot: 1:1000;

Type: Primary
Antigen: EEF1A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Chicken, Mouse, Non-Human Primate, Rat, Sheep, Xenopus

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty