Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ESR1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BB9
10802-630EA 504 USD InStock
10802-630
Anti-ESR1 Rabbit Polyclonal Antibody
Antibodies
Estrogen receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of ligand-activated transcription factors (1). Estrogen receptors, including ESR1, also known as ER-alpha and ESR2 (ER-beta), contain DNA binding and ligand binding domains and are critically involved in regulating the normal function of reproductive tissues. ESR1 is a widely expressed nuclear protein and serves as a strong activator of estrogen responsive genes (1,2). Phosphorylation of serines 104 and 106, located in the N-terminal transcription activation function-1 domain (AF-1), plays a large role in regulating ER alpha activity (3).

Recommended Dilutions: ELISA: 1:10,000-1:20,000; Immunohistochemsitry: User Optimized; Immunofluorescence Microscopy: User Optimized; Western Blot: 1-2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: ESR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty