Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FAM193B Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-BD3
10802-746EA 518 USD InStock
10802-746
Anti-FAM193B Rabbit Polyclonal Antibody
Antibodies
FAM193B, also known as IRIZIO, was initially identified as a protein that is upregulated in alveolar rhabdomyosarcoma (ARMS), a type of fast-growing tumor characterized by chromosomal translocations fusing the PAX3 or PAX7 gene with that of FOXO1. It has been suggested that, in addition to the PAX3-FOXO1 gene fusion, disruption of the Rb and p53 pathways is required for full ARMS tumorigenesis. In PAX3-FOXO1-expressing primary mouse fibroblasts that possessed a defective p53 pathway, FAM193B expression enabled the tumorigenic transformation, suggesting that FAM193B may contribute to rhabdomyosarcomagenesis in humans.

Recommended Dilutions: ELISA: 1:10,000; Immunofluorescence Microscopy: 20 ?g/mL; Western Blot: 1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: FAM193B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty