Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FGF4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-P57
10802-784EA 487 USD InStock
10802-784
Anti-FGF4 Rabbit Polyclonal Antibody
Antibodies
Fibroblast growth factor 4 (FGF4), also known as Heparin Secretary Transforming (HSTF1). HST1, for which the designation HSTF1 was proposed for human gene nomenclature, is a heparin-binding growth factor with significant homology to human fibroblast growth factors and the mouse Int-2 protein. By in situ hybridization, Adelaide et al. (1988) mapped the HST gene to chromosome 11q13. The HST1 protein is a heparin-binding growth factor with significant homology with human fibroblast growth factors and the mouse Int-2 protein. This antibody is suitable for researchers interested in cell signaling and cancer research.

Recommended Dilutions: Western Blot: 0.5µg/mL; 0.05mg Thimerosal

Type: Primary
Antigen: FGF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Internal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty