Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Mouse Monoclonal Antibody [clone: 8B9.C6.D3]

Supplier: Rockland Immunochemicals
200-301-096L
10802-928EA 2247 USD InStock
10802-928
Anti-IgG Mouse Monoclonal Antibody [clone: 8B9.C6.D3]
Antibodies
Anti-Fluorescein is designed for immunofluorescence microscopy, in situ hybridization, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.

Recommended Dilutions: ELISA: 1:5,000 - 1:30,000; Flow Cytometry: 1-2 µg/mL; Immunohistochemsitry: 1:250 - 1:1,500; Western Blot: 1:500 - 1:3,000; contains 0.01% (w/v) Sodium Azide

Type: Primary
Antigen: IgG
Clonality: Monoclonal
Clone: 8B9.C6.D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty