Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RBFOX3 Mouse Monoclonal Antibody [clone: 1B7]

Supplier: Rockland Immunochemicals
200-301-X09
10802-950EA 528 USD InStock
10802-950
Anti-RBFOX3 Mouse Monoclonal Antibody [clone: 1B7]
Antibodies
FOX3, also known as NeuN and hexaribonucleotide binding protein 3, is a neuron-specific RNA binding nuclear protein involved in the regulation of pre-mRNA alternative splicing. FOX3 dependent alternative splicing of Numb has recently been shown to play an important role in the progression of neuronal differentiation during vertebrate development. FOX3 Antibody is ideal for researchers interested in neuroscience research.

Recommended Dilutions: ELISA: 1:20,000; Immunofluorescence Microscopy: 1:500; Western Blot: 1:2000; 0.1% (w/v) Sodium Azide

Type: Primary
Antigen: RBFOX3
Clonality: Monoclonal
Clone: 1B7
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Mouse
Isotype:
Reactivity: Human, Bovine, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty