Would you like to log in again?
96-Well C8 Single-Break LUMITRAC™ 600 Strip Plate, Greiner Bio-One
Supplier: Greiner bio-one
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
705092
10803-076CS 787.5 USD InStock
10803-076
96-Well C8 Single-Break LUMITRAC™ 600 Strip Plate, Greiner Bio-One
Microplates Immunology Microplates and Strips
Break-apart strips with frame.
  • PS Material
  • High binding
C-bottom (flat with curved edge) strips. White wells.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty