Would you like to log in again?
VWR® Disposable Weighing Boats
Supplier: Avantor
VWR®
HS1424AAV HS1424AV HS1424CV HS1426AV HS1425CV HS1426AAV HS1425AV HS1427BV HS1426BV HS1425BV HS1427CV HS1426CV HS1427AV HS1425AAV HS1424BV HS1427AAV
10803-132CS 3123.73 USD InStock
10803-132 10803-110 10803-120 10803-142 10803-130 10803-114 10803-158 10803-124 10803-156 10803-112 10803-134 10803-122 10803-144 10803-118 10803-128 10803-126
VWR® Disposable Weighing Boats
Boats Weighing Boats
These polystyrene, diamond-shaped flexible boats feature smooth surfaces to provide accurate pour-out with minimal sample loss.
  • Wide, flat bottom avoids tipping
  • Temperature resistant up to +70°C
Boats are ideal for use with microwaves and moisture analysers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty