Would you like to log in again?
VWR® Disposable Square Weighing Boats
Supplier: VWR
VWR®
HS1423CV HS1420AV HS1422V HS1420CCV HS120304 HS1420CV HS1421AV HS1423BBV HS1423CCV HS1423AV HS120302 HS1423BV HS1420BBV HS120303 HS1420AA HS1423AAV HS1421CV HS1421BV HS1420BV
10803-172CS 393.42 USD InStock
10803-172 10770-448 10803-150 10803-170 10770-406 10803-154 10803-176 10803-164 10803-152 10803-174 10803-162 10770-442 10770-450 10803-136 10803-146 10803-168 10803-116 10803-138 10803-148
VWR® Disposable Square Weighing Boats
Boats Weighing Boats
These square shaped polystyrene flexible weighing boats with smooth surfaces enable accurate pour-out with minimal sample loss.
  • Shallow form with wide, flat bottom avoids tipping
  • Rounded corners simplify transfer
  • Temperature resistant up to 70°C
They are biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bleaches. Ideal for use with microwaves, balances, or moisture analyzers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty