Would you like to log in again?
Open-End Wrench Set, Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
20387
10803-216EA 171.19 USD InStock
10803-216
Open-End Wrench Set, Restek
Wrenches
High-quality 1/4 x 5/16", 3/8 x 7/16", 7/16 x 1/2", and 1/2 x 9/16" wrenches for tightening a wide variety of chromatography fittings.
  • Durable fabrication
  • Open-ended wrenches allow for easy application to fittings
  • Range of sizings
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty