Would you like to log in again?
MXT®-Union Connector Kits for Connecting Metal and/or Fused Silica GC Columns, Restek
Supplier: Restek
MXT®
21384 21386 21385
10852-942EA 183.53 USD InStock
10852-942 10852-944 10803-244
MXT®-Union Connector Kits for Connecting Metal and/or Fused Silica GC Columns, Restek
Fittings and Connectors
Connector kits for metal or fused silica columns are ideal for connecting guard columns or transfer lines to analytical columns.
  • Low dead volume, leak-tight connection
  • Reusable
  • Siltek® treatment ensures maximum inertness
  • Ideal for connecting a guard column or transfer line to an analytical column
  • Use to oven temperatures of 350°C
  • Available in union and “Y” configurations
  • Can also be used for connecting fused silica to metal
Each kit contains the MXT® union; two 1/32-inch nuts; and two, one-piece, fused silica adapters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty